Learn More
Medchemexpress LLC Glycine-15N | 7299-33-4 | MFCD00064426 | 99.8% | 76.06 | C2H5(15)NO2 | 50mg
SDP
Catalog No. 5000267663
Encompass_Preferred
Glycine-15N is the 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors[1]
Catalog No. 50-002-67663 Supplier Medchemexpress LLC Supplier No. HYY0966S50MG
Glycine-15N is the 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors[1]