Learn More
Medchemexpress LLC Glycine-2-13C,15N | 91795-59-4 | 77.05 | C13CH515NO2 | 25mg
SDP
Catalog No. 5000267734
Encompass_Preferred
Glycine-2-13C 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-67734 Supplier Medchemexpress LLC Supplier No. HYY0966S725MG
Glycine-2-13C 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors