Learn More
Medchemexpress LLC Glycine-2-13C | 20220-62-6 | MFCD00037550 | 99.3% | 76.06 | C13CH5NO2 | 10mg
SDP
Catalog No. 5000267827
Encompass_Preferred
Glycine-2-13C is the 13C-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-67827 Supplier Medchemexpress LLC Supplier No. HYY0966S210MG
Glycine-2-13C is the 13C-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors