Learn More
American Radiolabeled Chemicals Inc GLYCINE-2-3H 250UC
SDP
Catalog No. 5090107793
Encompass_Preferred
Glycin-2-3H 250uCi in ethanol:water (2:98), 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 75.1
Catalog No. 50-901-07793 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 0161A250UC
Glycin-2-3H 250uCi in ethanol:water (2:98), 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 75.1