Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycine, 56-40-6, MFCD00008131, 500 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G8898500G
Encompass
Linear Formula: NH2CH2COOH, Molecular Weight: 75.07, for electrophoresis, ≥99%, Synonym: Aminoacetic acid, Aminoethanoic acid, Glycocoll. Glycine has been used for the preparation of gelatin composites scaffold.
Catalog No. NC1167474