Learn More
Medchemexpress LLC HY-Y0966 500mg Medchemexpress, Glycine CAS:56-40-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0966500MG
Encompass_Preferred
Medchemexpress, HY-Y0966 500mg Glycine CAS:56-40-6 Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate, facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0768