Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycine BioUltra, for molecular biology | 56-40-6 | MFCD00008131 | 50g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 5004650G
Encompass
Glycine BioUltra, for molecular biology | Purity: 99% | Mol Wt: 75.07 | 56-40-6 | MFCD00008131 | 50g
Catalog No. NC1269857