Learn More
Bioworld Glycine, 500 g
Catalog No. 50256032
Encompass_Preferred
A reagent used in buffers.A compound used for polyacrylamide gel electrophoresis and Western blots Technical Information:Biotechnology GradeA280 (1.0M, water): <0.1Moisture: <0.2%Chromatography: Single spotHeavy metals (as Pb): <0.002%Appearance: White solidBoiling Point : ~240.90 °C at 760 mmHg (Predicted)Density : 1.1607 g/cm3HS Code : 292249100IUPAC_Name: GlycineMelting Point: 233 °CRTECS Number: MB7600000
Catalog No. 50-256-032 Supplier Bioworld Supplier No. 407000441
A reagent used in buffers.A compound used for polyacrylamide gel electrophoresis and Western blots Technical Information:Biotechnology GradeA280 (1.0M, water): <0.1Moisture: <0.2%Chromatography: Single spotHeavy metals (as Pb): <0.002%Appearance: White solidBoiling Point : ~240.90 °C at 760 mmHg (Predicted)Density : 1.1607 g/cm3HS Code : 292249100IUPAC_Name: GlycineMelting Point: 233 °CRTECS Number: MB7600000