Learn More
Medchemexpress LLC Glycine | 56-40-6 | MFCD00008131 | 99.9% | 75.07 | C2H5NO2 | 1g
SDP
Catalog No. 5000268138
Encompass_Preferred
Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors Glycine is orally active Glycine can be used to study cell protection cancer neurological diseases and angiogenesis[1][2][3][4][5][6]
Catalog No. 50-002-68138 Supplier Medchemexpress LLC Supplier No. HYY09661G
Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors Glycine is orally active Glycine can be used to study cell protection cancer neurological diseases and angiogenesis[1][2][3][4][5][6]