Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine benzyl ester hydrochloride, 97%

Catalog No. AC428540010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428540010 1 g Glass Bottle
AC428540050 5 g Glass Bottle
AC428540250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC428540010 Supplier Thermo Scientific Chemicals Supplier No. 428540010
Add to Cart

CAS: 2462-31-9 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2462-31-9
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035442
InChI Key VLQHNAMRWPQWNK-UHFFFAOYSA-N
Synonym benzyl glycinate hydrochloride, benzyl 2-aminoacetate hydrochloride, glycine benzyl ester hydrochloride, h-dl-gly-obzl.hcl, benzyl glycinate hcl, h-gly-obzl.hcl, benzyl glycinate, h-gly-obzl hcl, glycine benzyl ester hcl, glycine, phenylmethyl ester, hydrochloride
PubChem CID 11701227
IUPAC Name benzyl 2-aminoacetate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)CN.Cl

Specifications

Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 201.65
Percent Purity 97%
Chemical Name or Material Glycine benzyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.