Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycine Buffer Solution | 100ml
Catalog No. NC1737317
Encompass
Glycine is used as a bulking agent buffers. Glycine at low concentrations prevents pH decrease in solutions. It also stabilizes a protein when present in an amorphous state.
Catalog No. NC1737317 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G5418100ML
Glycine is used as a bulking agent buffers. Glycine at low concentrations prevents pH decrease in solutions. It also stabilizes a protein when present in an amorphous state.