Learn More
Medchemexpress LLC Glycine-d2 | 4896-75-7 | MFCD00075077 | 99.9% | 77.08 | C2H3D2NO2 | 500mg
SDP
Catalog No. 5000268145
Encompass_Preferred
Glycine-d2 is the deuterium labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-68145 Supplier Medchemexpress LLC Supplier No. HYY0966S1500MG
Glycine-d2 is the deuterium labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors