Learn More
Medchemexpress LLC Glycine-d5 | 4896-77-9 | MFCD00064427 | 98.0% | 80.10 g·mol⁻¹ | C2D5NO2 | 100mg
SDP
Catalog No. 5000268153
Encompass_Preferred
Glycine-d5 is the deuterium labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-68153 Supplier Medchemexpress LLC Supplier No. HYY0966S8100MG
Glycine-d5 is the deuterium labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors