Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, Electrophoresis Grade, MP Biomedicals™

Catalog No. ICN808831 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN808831 5 kg
ICN808822 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN808831 Supplier MP Biomedicals, Inc Supplier No. 04808831
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycine, Electrophoresis Grade is for use in electrophoresis buffers and SDS-PAGE.

  • Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis.
  • Glycine is also a component of Towbin′s transfer buffer for Western blots.
  • It is an inhibitory neurotransmitter in the spinal cord and an allosteric regulator of NMDA receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 451°F
pH 4 (0.2 molar aq. soln.)
Specific Rotation ≤0.0065%
Solubility Information Soluble in water. Slightly soluble in alcohol and ethanol (95%). Almost insoluble in ether.
Density 1.1607 g/mL
Quantity 5 kg
Optical Rotation ≤0.0065%
Percent Purity ≥99.5%
Grade Electrophoresis
Physical Form Powder
pKa 2.35 at 25°C
Chemical Name or Material Glycine
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.