Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603008 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-008 50 kg Fiber Drum
18-603-004 100 g Poly Bottle
18-603-005 500 g Poly Bottle
18-603-006 2.5 kg Poly Pail
18-603-007 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-008 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G104150KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G1041, 56-40-6, C2H5NO2

Spectrum™ Chemical Glycine, FCC is an organic compound and is also the smallest of the 20 common amino acids in proteins. It is a sweet tasting, colorless, crystalline solid. Glycine is a common building block to many natural products and is a precursor to proteins It is also used as an additive in animal food, and as a food supplement and sweetener in food destined for human consumption. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Lead (Pb) 5 mg/kg
Loss on Drying 0.2%
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 98.5 to 101.5%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.