Learn More
Research Products International Corp Glycine, Free Base, 2.5 Kilograms
SDP
Supplier:  Research Products International Corp G360502500.0
Encompass_Preferred
For use in preparing electrophoresis buffers. Cell culture tested.
Catalog No. 50-136-8284