Learn More
Research Products International Corp Glycine, Free Base, 5 Kilograms
SDP
Supplier:  Research Products International Corp G360505000.0
Encompass_Preferred
For use in preparing electrophoresis buffers. Cell culture tested.
Catalog No. 50-213-342