Learn More
Polysciences, Inc. 1M Glycine-HCl Buffer, pH 2.7+/-0.1, 10X Concentrate, 1 L
SDP
Catalog No. NC9554201
Encompass
Dilute 1-10 with deionized water to get buffer at working concentration.
Catalog No. NC9554201 Supplier Polysciences, Inc. Supplier No. 240741
Dilute 1-10 with deionized water to get buffer at working concentration.