Learn More
Aniara Diagnostica LLC Glycine-HCl, pH 3.0, 10 pouches
Catalog No. 501008832
Encompass
Medicago Glycine-HCl Buffer pH 3.0 supplied as preweighed powder mix in sealed pouch yields 1000 mL 0.1 M glycine-HCl buffer with pH 3.0 at 25C. Used for elution buffer in chromatography procedures. 10 pouches. Glycine-HCI is used as an elution buffer in Protein A chromatography and other types of affinity chromatography. Medicago's glycine is more than 99% pure.
Catalog No. 50-100-8832 Supplier Aniara Diagnostica LLC Supplier No. A12912110
Medicago Glycine-HCl Buffer pH 3.0 supplied as preweighed powder mix in sealed pouch yields 1000 mL 0.1 M glycine-HCl buffer with pH 3.0 at 25C. Used for elution buffer in chromatography procedures. 10 pouches. Glycine-HCI is used as an elution buffer in Protein A chromatography and other types of affinity chromatography. Medicago's glycine is more than 99% pure.