Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycine, 56-40-6, MFCD00008131, 1 kg
Catalog No. NC0756056
Encompass
Linear Formula: NH2CH2COOH, Molecular Weight: 75.07, ReagentPlus, ≥99% (HPLC), Synonym: Aminoacetic acid, Aminoethanoic acid, Glycocoll.
Catalog No. NC0756056 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G71261KG
Linear Formula: NH2CH2COOH, Molecular Weight: 75.07, ReagentPlus, ≥99% (HPLC), Synonym: Aminoacetic acid, Aminoethanoic acid, Glycocoll.