Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycine hydrochloride, 6000-43-7, MFCD00012872, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G2879100G
Encompass
Linear Formula: H2NCH2CO2H · HCl, Molecular Weight: 111.53, ≥99% (HPLC), mp: 176-180 °C, Synonym: Aminoacetic acid, Aminoethanoic acid, Glycocoll.
Catalog No. NC0618611