Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. G010325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G010325G Supplier TCI America Supplier No. G010325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent for research applications.

Chemical Identifiers

CAS 6000-43-7
Molecular Formula C2H6ClNO2
Molecular Weight (g/mol) 111.53
MDL Number MFCD00012872
InChI Key IVLXQGJVBGMLRR-UHFFFAOYSA-N
Synonym glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
PubChem CID 22316
IUPAC Name hydrogen 2-aminoacetic acid chloride
SMILES [H+].[Cl-].NCC(O)=O

Specifications

Melting Point 179°C
Color White
Quantity 25 g
Formula Weight 111.53
Percent Purity ≥99.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Glycine Hydrochloride
RTECSNumber MC0560000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.