Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycine Methyl Ester Hydrochloride

Catalog No. G6600500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G6600500G 100 g
G6600100G 5 g
G66005G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G6600500G Supplier Sigma Aldrich Supplier No. G6600500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycine Methyl Ester Hydrochloride

Specifications

CAS 5680-79-5
Melting Point 175°C
Molecular Formula C3H7NO2 -+ HCl
Linear Formula NH2CH2COOCH3 -+ HCl
MDL Number MFCD00012870
Quantity 100 g
Molecular Weight (g/mol) 125.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.