Learn More
Medchemexpress LLC Glycine, N-(1-oxooctadecyl)- | 6333-54-6 | MFCD00153472 | 100.0% | 341.53 g/mol | C20H39NO3 | 250 MG
SDP
Catalog No. 5000282966
Encompass_Preferred
N-Stearoylglycine is an N-acylglycine lipid with an 18-carbon stearoyl chain. It has an ionizable glycine headgroup whose charge depends on pH and an amide linkage capable of forming hydrogen-bonded networks in ordered films, making it suitable for studies of lipid assembly, model membranes, and surface interactions.
  • Useful for model membrane and lipid self-assembly studies.
  • Ionizable headgroup for pH-dependent behavior and surface chemistry experiments.
  • Amide moiety supports hydrogen-bonded network formation in ordered films.
  • High purity analytical grade (99.95%).
  • Molecular weight 341.53 g/mol; formula C20H39NO3.
Catalog No. 50-002-82966 Supplier Medchemexpress LLC Supplier No. HYW141932250MG
N-Stearoylglycine is an N-acylglycine lipid with an 18-carbon stearoyl chain. It has an ionizable glycine headgroup whose charge depends on pH and an amide linkage capable of forming hydrogen-bonded networks in ordered films, making it suitable for studies of lipid assembly, model membranes, and surface interactions.
  • Useful for model membrane and lipid self-assembly studies.
  • Ionizable headgroup for pH-dependent behavior and surface chemistry experiments.
  • Amide moiety supports hydrogen-bonded network formation in ordered films.
  • High purity analytical grade (99.95%).
  • Molecular weight 341.53 g/mol; formula C20H39NO3.