Learn More
Medchemexpress LLC Glycine, N-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]- | 2412056-27-8 | 95.0% | C15H13N3O6 | 25 MG
SDP
Catalog No. 5000358926
Encompass_Preferred
Thalidomide-5-NH2-CH2-COOH (compound 114) is a potent and selective inhibitor of tropomyosin receptor kinase (trk) and functions as a ligand of E3 ligase. This PROTAC leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins, holding potential for researching various diseases, as extracted from patent WO2021170109A1. This product is for research use only.
  • Potent and selective inhibitor of tropomyosin receptor kinase (trk)
  • Functions as a ligand for E3 ligase
  • PROTAC (proteolysis-targeting chimera) technology
  • Utilizes the ubiquitin-proteasome system to degrade target proteins
  • Potential for researching various diseases
Catalog No. 50-003-58926 Supplier Medchemexpress LLC Supplier No. 25MGHY46531
Thalidomide-5-NH2-CH2-COOH (compound 114) is a potent and selective inhibitor of tropomyosin receptor kinase (trk) and functions as a ligand of E3 ligase. This PROTAC leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins, holding potential for researching various diseases, as extracted from patent WO2021170109A1. This product is for research use only.
  • Potent and selective inhibitor of tropomyosin receptor kinase (trk)
  • Functions as a ligand for E3 ligase
  • PROTAC (proteolysis-targeting chimera) technology
  • Utilizes the ubiquitin-proteasome system to degrade target proteins
  • Potential for researching various diseases