Learn More
Medchemexpress LLC Glycine, N-(2-amino-2-oxoethyl)-N-(carboxymethyl)-, sodium salt (1:2) | 41689-31-0 | 98.0% | 25 G
SDP
Catalog No. 5000414094
Encompass_Preferred
ADA disodium is a buffer salt derived from ADA (adenosine deaminase) and is widely used in biochemical research, especially in the preparation of enzyme buffers.
  • Derived from adenosine deaminase
  • Used in biochemical research
  • Ideal for enzyme buffer preparation
Catalog No. 50-004-14094 Supplier Medchemexpress LLC Supplier No. 25GHYD0855A
ADA disodium is a buffer salt derived from ADA (adenosine deaminase) and is widely used in biochemical research, especially in the preparation of enzyme buffers.
  • Derived from adenosine deaminase
  • Used in biochemical research
  • Ideal for enzyme buffer preparation