Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycine, N-(2-amino-2-oxoethyl)-N-(carboxymethyl)-, sodium salt (1:2) | 41689-31-0 | 98.0% | 50 G
SDP

Catalog No. 5000410164
Encompass_Preferred

ADA disodium is a buffer salt derived from adenosine deaminase, commonly used in biochemical research for preparing enzyme buffers. This product is intended for research use only and is not for human or animal therapeutic use.

  • Solid appearance
  • White to off-white color
  • Molecular weight: 234.12
  • Chemical formula: C6H8N2Na2O5
  • Solubility in water: 350 mg/mL
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture

Catalog No. 50-004-10164 Supplier Medchemexpress LLC Supplier No. HYD0855A50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ADA disodium is a buffer salt derived from adenosine deaminase, commonly used in biochemical research for preparing enzyme buffers. This product is intended for research use only and is not for human or animal therapeutic use.

  • Solid appearance
  • White to off-white color
  • Molecular weight: 234.12
  • Chemical formula: C6H8N2Na2O5
  • Solubility in water: 350 mg/mL
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.