Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycine, N-(3-hydroxybenzoyl)- | 1637-75-8 | 99.1% | 195.17 | 10 MG
SDP

Catalog No. 5000435943
Encompass_Preferred

3-Hydroxyhippuric acid (Glycine, N-(3-hydroxybenzoyl)-) is a chemical compound supplied as a white to off-white solid. It has a molecular weight of 195.17 and a purity of 99.09% as determined by LCMS. This product is intended for research use only and has not been fully validated for medical applications.

  • White to off-white solid appearance
  • Molecular formula: C9H9NO4
  • Purity: 99.09% (LCMS)
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 2 years, -20°C for 1 year
  • For research use only

Catalog No. 50-004-35943 Supplier Medchemexpress LLC Supplier No. HY113085R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxyhippuric acid (Glycine, N-(3-hydroxybenzoyl)-) is a chemical compound supplied as a white to off-white solid. It has a molecular weight of 195.17 and a purity of 99.09% as determined by LCMS. This product is intended for research use only and has not been fully validated for medical applications.

  • White to off-white solid appearance
  • Molecular formula: C9H9NO4
  • Purity: 99.09% (LCMS)
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 2 years, -20°C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.