Learn More
Medchemexpress LLC Glycine, N-[(3R)-3-([1,1'-biphenyl]-4-yloxy)-3-(4-fluorophenyl)propyl]-N-methyl-, hydrochloride | 200006-08-2 | MFCD09971056 | 98.2% | 429.91 g/mol | C24H25ClFNO3 | 5 MG
SDP
Catalog No. 5000239904
Encompass_Preferred
ALX-5407 hydrochloride is the hydrochloride salt of a research-grade small-molecule inhibitor of the glycine transporter GlyT1. It is used to probe glycine uptake and NMDA receptor modulation in preclinical studies and is supplied as a characterized solid with high purity and documented handling conditions.
  • Potent GlyT1 inhibitor with reported IC50 ≈ 3 nM.
  • Selective for GlyT1 over GlyT2 and NMDA receptors.
  • High purity solid suitable for research applications.
  • Characterized molecular weight and formula for reproducibility.
  • Recommended sealed storage at 4°C; solvent storage conditions documented.
Catalog No. 50-002-39904 Supplier Medchemexpress LLC Supplier No. HY10711A5MG
ALX-5407 hydrochloride is the hydrochloride salt of a research-grade small-molecule inhibitor of the glycine transporter GlyT1. It is used to probe glycine uptake and NMDA receptor modulation in preclinical studies and is supplied as a characterized solid with high purity and documented handling conditions.
  • Potent GlyT1 inhibitor with reported IC50 ≈ 3 nM.
  • Selective for GlyT1 over GlyT2 and NMDA receptors.
  • High purity solid suitable for research applications.
  • Characterized molecular weight and formula for reproducibility.
  • Recommended sealed storage at 4°C; solvent storage conditions documented.