Learn More
Medchemexpress LLC Glycine, N,N-bis[2-(2,6-dioxo-4-morpholinyl)ethyl]- | 23911-26-4 | ≥97.0% | 357.32 | 10 G
SDP
Catalog No. 5000451798
Encompass_Preferred
DTPA anhydride is a bifunctional chelator. Its anhydride can react with amino groups in proteins, such as lysine residues, to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs). Radionuclide-labeled drug conjugates have the ability to specifically target biomolecules and can be used in medical imaging or therapy. This product is for research use only.
  • Bifunctional chelator
  • Reacts with amino groups in proteins
  • Forms stable amide bonds
  • Binds to radionuclides
  • Used to synthesize radionuclide-labeled drug conjugates (RDCs)
  • Radionuclide-labeled drug conjugates can be used in medical imaging or therapy
  • For research use only
Catalog No. 50-004-51798 Supplier Medchemexpress LLC Supplier No. HYW01111810G
DTPA anhydride is a bifunctional chelator. Its anhydride can react with amino groups in proteins, such as lysine residues, to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs). Radionuclide-labeled drug conjugates have the ability to specifically target biomolecules and can be used in medical imaging or therapy. This product is for research use only.
  • Bifunctional chelator
  • Reacts with amino groups in proteins
  • Forms stable amide bonds
  • Binds to radionuclides
  • Used to synthesize radionuclide-labeled drug conjugates (RDCs)
  • Radionuclide-labeled drug conjugates can be used in medical imaging or therapy
  • For research use only