Learn More
Medchemexpress LLC Glycine, N,N-bis[2-(2,6-dioxo-4-morpholinyl)ethyl]- | 23911-26-4 | ≥97.0% | 357.32 | 25 G
SDP
Catalog No. 5000366890
Encompass_Preferred
DTPA anhydride is a bifunctional chelator whose anhydride can react with amino groups in proteins (such as lysine residues) to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs). RDCs have the ability to specifically target biomolecules and can be used in medical imaging or therapy.
  • Bifunctional chelator
  • Reacts with amino groups in proteins
  • Forms stable amide bonds
  • Binds to radionuclides
  • Synthesizes radionuclide-labeled drug conjugates (RDCs)
  • RDCs can specifically target biomolecules
  • Used in medical imaging or therapy
Catalog No. 50-003-66890 Supplier Medchemexpress LLC Supplier No. 25GHYW011118
DTPA anhydride is a bifunctional chelator whose anhydride can react with amino groups in proteins (such as lysine residues) to form stable amide bonds. It can also bind to radionuclides to synthesize radionuclide-labeled drug conjugates (RDCs). RDCs have the ability to specifically target biomolecules and can be used in medical imaging or therapy.
  • Bifunctional chelator
  • Reacts with amino groups in proteins
  • Forms stable amide bonds
  • Binds to radionuclides
  • Synthesizes radionuclide-labeled drug conjugates (RDCs)
  • RDCs can specifically target biomolecules
  • Used in medical imaging or therapy