Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycine, N-(phenylmethyl)-, ethyl ester | 6436-90-4 | 99.1% | 193.25 | 500 G
SDP

Catalog No. 5000453162
Encompass_Preferred

N-Benzylglycine ethyl ester is an N-substituted glycine derivative that is bound to PDB code: 1B41 and can be used in Alzheimer's disease research. This compound appears as a colorless to light yellow liquid.

  • N-substituted glycine derivative
  • Used in Alzheimer's disease research
  • Purity of 99.09% by GC
  • Pure form storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-004-53162 Supplier Medchemexpress LLC Supplier No. HYW009970500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzylglycine ethyl ester is an N-substituted glycine derivative that is bound to PDB code: 1B41 and can be used in Alzheimer's disease research. This compound appears as a colorless to light yellow liquid.

  • N-substituted glycine derivative
  • Used in Alzheimer's disease research
  • Purity of 99.09% by GC
  • Pure form storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.