Learn More
Hach Company Glycine Reagent, 100 mL
Supplier:  Hach Company 2344042
Encompass
Reagent for manganese interference correction in Indigo Method for ozone. 100 mL Bottle
Catalog No. 50-199-5837