Learn More
Enzo Life Sciences Glycine receptor polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA5100200
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of N-terminal rat glycine receptor subunit α1. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Formulation: Lyophilized from 5mM ammonium bicarbonate.
Catalog No. 50-201-1843