Learn More
Bioworld GLYCINE SEPAROPOREAGAROSE
Supplier:  Bioworld 201810653
Encompass
GLYCINE SEPAROPORE (AGAROSE) 4B: Specifications: Ligand: Glycine.; Particle size (range): 50-175m.; Molecular Weight range: 6x104 - 2x107.; Matrix: Separopore 4B (Agarose 4% Beads).; pH stability: 4-9.; Flow specifications: 70-140 cm/h.;...
Catalog No. 50-197-833