Learn More
Accela Chembio Inc MF: C6H14ClNO2 | MW: 167.63 | >95% | Flash Point: 20.7? | BP: 145.7? at 760 mmHg.
Supplier:  Accela Chembio Inc SY003945100G
Encompass_Preferred
MF: C6H14ClNO2 | MW: 167.63 | >95% | Flash Point: 20.7? | BP: 145.7? at 760 mmHg.
Catalog No. 50-353-924