Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603010 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-010 500 g Poly Bottle
18-603-011 2.5 kg Poly Bottle
18-603-012 12 kg Poly Pail
18-603-013 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-010 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AM125500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

AM125, 56-40-6, C2H5NO2

Spectrum™ Chemical Glycine, USP is a colorless, sweet-tasting crystalline non-essential amino acid. USP glycine has a wide variety of uses, including as an additive in pet food and animal feed, in foods and pharmaceuticals as a sweetener/taste enhancer, or as a component of food supplements and protein drinks. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

CAS Max % 100%
Chloride 0.007%
Ignition Residue 0.1%
Loss on Drying 0.2%
Packaging Poly Bottle
Sulfate 0.0065%
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.