Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine USP/EP/JP/FCC, Ajinomoto

Catalog No. 502946066
Change view
Click to view available options
Packaging:
Fiber Drum
Poly Drum
Quantity:
1 kg
10 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
50-294-6066 Poly Drum 50 kg
50-294-6082 Fiber Drum 50 kg
50-294-5989 Poly Drum 1 kg
50-294-5979 Poly Drum 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-294-6066 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A33486
Only null left
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 56-40-6
Molecular Weight (g/mol) 75.07

Specifications

Packaging Poly Drum
Quantity 50 kg
Percent Purity 98.5 to 101.0%
Chemical Name or Material Glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.