Learn More
Gold Biotechnology Inc Glycine USP Grade 5 kg
SDP
Catalog No. 5000562076
Encompass_Preferred
Because of its charge properties and small size glycine is commonly used as a counter-ion in SDS-PAGE applications to balance running buffer charge and electric field strength
Catalog No. 50-005-62076 Supplier Gold Biotechnology Inc Supplier No. G6305
Because of its charge properties and small size glycine is commonly used as a counter-ion in SDS-PAGE applications to balance running buffer charge and electric field strength