Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycinexylidide | 18865-38-8 | MFCD05267700 | 95.1% | 178.23 g/mol | C10H14N2O | 250 MG
SDP

Catalog No. 5000285195
Encompass_Preferred

Glycinexylidide is the active metabolite of lidocaine supplied as a research reagent for biochemical, pharmacokinetic, and in vitro pharmacology studies. It is used as a reference standard and for investigations into lidocaine metabolism and sodium channel-related activity.

  • Active metabolite of lidocaine used in metabolism and pharmacokinetic studies.
  • Supplied as a research-grade chemical reagent for laboratory use.
  • Available in small analytical quantities suitable for assay and reference standard use.
  • Purity around 95.1%, supporting reproducible experimental results.
  • Molecular formula C10H14N2O and molecular weight 178.23 g/mol.

Catalog No. 50-002-85195 Supplier Medchemexpress LLC Supplier No. HYW357818250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycinexylidide is the active metabolite of lidocaine supplied as a research reagent for biochemical, pharmacokinetic, and in vitro pharmacology studies. It is used as a reference standard and for investigations into lidocaine metabolism and sodium channel-related activity.

  • Active metabolite of lidocaine used in metabolism and pharmacokinetic studies.
  • Supplied as a research-grade chemical reagent for laboratory use.
  • Available in small analytical quantities suitable for assay and reference standard use.
  • Purity around 95.1%, supporting reproducible experimental results.
  • Molecular formula C10H14N2O and molecular weight 178.23 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.