Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycitein

Catalog No. AC464642500
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464642500 250 mg
AC464640500 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464642500 Supplier Thermo Scientific Chemicals Supplier No. 464642500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 40957-83-3 | C16H12O5 | 284.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40957-83-3
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.27
InChI Key DXYUAIFZCFRPTH-UHFFFAOYSA-N
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one
SMILES COC1=C(O)C=C2OC=C(C(=O)C2=C1)C1=CC=C(O)C=C1

Specifications

Quantity 250 mg
Formula Weight 284.26
Chemical Name or Material Glycitein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.