Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avanti Polar Lipids Glyco-diosgenin, Powder, 5 g, Assay >99%, C56H92O25, CAS 1402423-29-3, Formal Structure, -20°C Storage, Shipped Dry Ice, 30 mL Amber Wide Mouth Bottle

Catalog No. NC3780549
Encompass

Glyco-diosgenin (GDN) is a high-purity amphiphile powder designed for scientific applications focused on the solubilization and stabilization of membrane proteins. Its distinctive structure, which includes rigid hydrophobic groups, contributes to its effectiveness in membrane protein research. The product comes in a 5g quantity, securely packaged in a 30 mL amber glass bottle to enhance stability and ease of handling.

  • Synonym: GDN
  • Empirical formula: C56H92O25
  • CAS Number: 1402423-29-3
  • Molecular weight: 1165.31
  • Form: Powder
  • Assay: >99% (TLC)
  • Quantity: 5 g
  • Storage temperature: -20°C
  • Shipped in: Dry ice
  • Packaging: 30 mL amber wide mouth screw cap glass bottle
  • UNSPSC Code: 12161902
  • Storage Class Code: 11 - Combustible solids
  • WGK: WGK 3

Catalog No. NC3780549 Supplier Avanti Polar Lipids Supplier No. 850525P5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glyco-diosgenin (GDN) is a high-purity amphiphile powder designed for scientific applications focused on the solubilization and stabilization of membrane proteins. Its distinctive structure, which includes rigid hydrophobic groups, contributes to its effectiveness in membrane protein research. The product comes in a 5g quantity, securely packaged in a 30 mL amber glass bottle to enhance stability and ease of handling.

  • Synonym: GDN
  • Empirical formula: C56H92O25
  • CAS Number: 1402423-29-3
  • Molecular weight: 1165.31
  • Form: Powder
  • Assay: >99% (TLC)
  • Quantity: 5 g
  • Storage temperature: -20°C
  • Shipped in: Dry ice
  • Packaging: 30 mL amber wide mouth screw cap glass bottle
  • UNSPSC Code: 12161902
  • Storage Class Code: 11 - Combustible solids
  • WGK: WGK 3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.