Learn More
Innovative Research Inc Human Glyco IIb/IIIa Purified 1mg
Supplier:  Innovative Research Inc IHUITGA2B1MG
Encompass_Preferred
Human Glycoprotein IIb/IIIa Purified from Innovative Research has been purified from Human Platelets. This is a frozen liquid buffered in 20 mM Tris-HCl/ 0.1 M NaCl/ 0.1% Triton X-100/ 1 mM CaCI2/ 0.05% NaN3/ 50% glycerol/ pH 7.4, with a concentration of 1mg/ml. As observed on gels, GPIIb migrates as 136,000 (non-reduced) and 125,000 (reduced), and GPIIIa as 97,000 (non-reduced) and 108,000 (reduced).
Glycoprotein IIb/IIIa is an integrin complex found on platelets that aids in platelet activation by functioning as a receptor for both Factor I (Fibrinogen) and von Willebrand Factor. The Glycoprotein IIb/IIIa complex is formed by calcium-dependent association of Glycoprotein IIb and Glycoprotien IIIa. This complex binds Factor I (Fibrinogen) to platelets, allowing multiple platelets to join together as they attach to the same strands of fibrinogen and form a clot. The coagulation cascade then continues, stabilizing the clot as Factor IIa (Thrombin) converts the soluble fibrino
Catalog No. 50-203-6570
 Restricted Item