Learn More
Vector Laboratories Glycoprotein Eluting Solution for Mannose/Glucose Binding Lectins | 100mL
SDP
Catalog No. NC1777776
Encompass
Vector Laboratories has developed Glycoprotein Elution Solutions in the neutral pH range that maximize the yield of eluted glycoproteins and preserve the activity of the agarose-bound lectins for repeated use. All components of these ready-to-use Glycoprotein Eluting Solutions can be subsequently removed by dialysis. These Glycoprotein Eluting Solutions offer researchers convenience and superior recovery over standard sugar solutions.
Catalog No. NC1777776 Supplier Vector Laboratories Supplier No. ES1100100
Vector Laboratories has developed Glycoprotein Elution Solutions in the neutral pH range that maximize the yield of eluted glycoproteins and preserve the activity of the agarose-bound lectins for repeated use. All components of these ready-to-use Glycoprotein Eluting Solutions can be subsequently removed by dialysis. These Glycoprotein Eluting Solutions offer researchers convenience and superior recovery over standard sugar solutions.