Learn More
Medchemexpress LLC HY-W001083 100mg Medchemexpress, 3-Hydroxyphenylacetic acid CAS:621-37-4 Purity:>98%
SDP
Catalog No. 501970633
Encompass_Preferred
Medchemexpress, HY-N2334A 100mg Glycochenodeoxycholic acid (sodium salt) CAS:16564-43-5 Glycochenodeoxycholic acid sodium salt (Chenodeoxycholylglycine sodium salt) is a bile acid formed in the liver from chenodeoxycholate and glycine. It acts as a detergent to solubilize fats for absorption and is itself absorbed. Glycochenodeoxycholic acid sodium salt (Chenodeoxycholylglycine sodium salt) induces hepatocyte apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0633 Supplier Medchemexpress LLC Supplier No. HYW001083100MG
Medchemexpress, HY-N2334A 100mg Glycochenodeoxycholic acid (sodium salt) CAS:16564-43-5 Glycochenodeoxycholic acid sodium salt (Chenodeoxycholylglycine sodium salt) is a bile acid formed in the liver from chenodeoxycholate and glycine. It acts as a detergent to solubilize fats for absorption and is itself absorbed. Glycochenodeoxycholic acid sodium salt (Chenodeoxycholylglycine sodium salt) induces hepatocyte apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.