Learn More
Cayman Chemical Glycochenodeoxycholic Acid (sodium salt hydrate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502965161
Encompass_Preferred
A glycine-conjugated form of chenodeoxycholic acid; reduces formation of cholic acid in primary human hepatocytes at 100 µM; reduces the number LC3 puncta and is cytotoxic to L-02 hepatocytes at 50, 75, and 100 µM; induces apoptosis in isolated rat hepatocytes at 50 µM; fecal levels are decreased in a rat model of high-fat diet-induced obesity compared with rats fed a normal diet
Catalog No. 50-296-5161 Supplier Cayman Chemical Supplier No. 1694250 MG
A glycine-conjugated form of chenodeoxycholic acid; reduces formation of cholic acid in primary human hepatocytes at 100 µM; reduces the number LC3 puncta and is cytotoxic to L-02 hepatocytes at 50, 75, and 100 µM; induces apoptosis in isolated rat hepatocytes at 50 µM; fecal levels are decreased in a rat model of high-fat diet-induced obesity compared with rats fed a normal diet