Learn More
Selleck Chemical LLC Glycochenodeoxycholic acid
SDP
Catalog No. 5000002949
Encompass_Preferred
Glycochenodeoxycholic acid is a bile salt formed in the liver from chenodeoxycholic acid and glycine usually found as the sodium salt It acts as a detergent to solubilize fats for absorption and is itself absorbed
Catalog No. 50-000-02949 Supplier Selleck Chemical LLC Supplier No. S635525MG
Glycochenodeoxycholic acid is a bile salt formed in the liver from chenodeoxycholic acid and glycine usually found as the sodium salt It acts as a detergent to solubilize fats for absorption and is itself absorbed