Learn More
Medchemexpress LLC Glycochenodeoxycholic acid-d4 | 1201918-16-2 | 1 MG
SDP
Catalog No. 5000420414
Encompass_Preferred
Glycochenodeoxycholic acid-d4 is the deuterium labeled Glycochenodeoxycholic acid. It is a bile acid formed in the liver from chenodeoxycholate and glycine, acting as a detergent to solubilize fats for absorption. This compound induces hepatocyte apoptosis.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-20414 Supplier Medchemexpress LLC Supplier No. HYN2334S1MG
Glycochenodeoxycholic acid-d4 is the deuterium labeled Glycochenodeoxycholic acid. It is a bile acid formed in the liver from chenodeoxycholate and glycine, acting as a detergent to solubilize fats for absorption. This compound induces hepatocyte apoptosis.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.