Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycochenodeoxycholic acid-d4 | 1201918-16-2 | MFCD00271498 | 99.9% | 453.65 g/mol | C26H39D4NO5 | 10 MG
SDP

Catalog No. 5000289921
Encompass_Preferred

Glycochenodeoxycholic acid-d4 is the deuterium-labeled analogue of glycochenodeoxycholic acid supplied as a high-purity analytical standard for mass spectrometry, metabolic tracing, and biochemical assays. It is a bile acid derivative that functions as a detergent to solubilize lipids and has been reported to induce hepatocyte apoptosis.

  • High isotopic purity for internal standard and tracer applications.
  • Purity 99.9% ensures reliable analytical sensitivity.
  • Molecular weight 453.65 g/mol and formula C26H39D4NO5.
  • Available in small vial sizes suitable for analytical workflows, including 10 mg.
  • Suitable for LC-MS/MS quantification and metabolic studies.
  • Provided with batch-specific datasheet and certificate of analysis.

Catalog No. 50-002-89921 Supplier Medchemexpress LLC Supplier No. HYN2334S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycochenodeoxycholic acid-d4 is the deuterium-labeled analogue of glycochenodeoxycholic acid supplied as a high-purity analytical standard for mass spectrometry, metabolic tracing, and biochemical assays. It is a bile acid derivative that functions as a detergent to solubilize lipids and has been reported to induce hepatocyte apoptosis.

  • High isotopic purity for internal standard and tracer applications.
  • Purity 99.9% ensures reliable analytical sensitivity.
  • Molecular weight 453.65 g/mol and formula C26H39D4NO5.
  • Available in small vial sizes suitable for analytical workflows, including 10 mg.
  • Suitable for LC-MS/MS quantification and metabolic studies.
  • Provided with batch-specific datasheet and certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.