Learn More
Medchemexpress LLC Glycochenodeoxycholic acid-d4 (Chenodeoxycholylglycine-d4) | 1201918-16-2 | 5 MG
SDP
Catalog No. 5000418982
Encompass_Preferred
Glycochenodeoxycholic acid-d4 is the deuterium labeled Glycochenodeoxycholic acid. Glycochenodeoxycholic acid (Chenodeoxycholylglycine) is a bile acid formed in the liver from chenodeoxycholate and glycine. It acts as a detergent to solubilize fats for absorption and is itself absorbed. Glycochenodeoxycholic acid (Chenodeoxycholylglycine) induces hepatocyte apoptosis.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can affect the pharmacokinetic and metabolic profiles of drugs.
  • Purity: 99.95%.
Catalog No. 50-004-18982 Supplier Medchemexpress LLC Supplier No. HYN2334S5MG
Glycochenodeoxycholic acid-d4 is the deuterium labeled Glycochenodeoxycholic acid. Glycochenodeoxycholic acid (Chenodeoxycholylglycine) is a bile acid formed in the liver from chenodeoxycholate and glycine. It acts as a detergent to solubilize fats for absorption and is itself absorbed. Glycochenodeoxycholic acid (Chenodeoxycholylglycine) induces hepatocyte apoptosis.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can affect the pharmacokinetic and metabolic profiles of drugs.
  • Purity: 99.95%.